ENA249152309 Enamine

ethyl 3-amino-3-phenylpropanoate hydrochloride

  • MDL number MFCD01902767
  • Cas Number : 29754-04-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 132...134
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-56135

Safety Information

msds https://www.enaminestore.com/catalog/EN300-56135
Availability details for
Quantity
  • Availability
  • Add to Cart