ENA249152313 Enamine

2-phenyl-2,3-dihydro-1H-isoindole-1,3-dione

  • MDL number MFCD00023044
  • Cas Number : 520-03-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 206-208
Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-56140

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-56140
Availability details for
Quantity
  • Availability
  • Add to Cart