ENA249152314 Enamine

5,6,7,8-tetrahydronaphthalen-1-ylboranediol

  • MDL number MFCD08701755
  • Cas Number : 371765-41-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 201...203
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-53921

Safety Information

msds https://www.enaminestore.com/catalog/EN300-53921
Availability details for
Quantity
  • Availability
  • Add to Cart