ENA249152316 Enamine

1-(3-benzyl-1,2,4-oxadiazol-5-yl)methanamine hydrochloride

  • MDL number MFCD09997606
  • Cas Number : 1185301-35-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-56143

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-56143
Availability details for
Quantity
  • Availability
  • Add to Cart