ENA249152317 Enamine

methyl 4-(2-methylphenyl)-2,4-dioxobutanoate

  • MDL number MFCD11188866
  • Cas Number : 1037130-77-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 59...61
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-56144

Safety Information

msds https://www.enaminestore.com/catalog/EN300-56144
Availability details for
Quantity
  • Availability
  • Add to Cart