ENA249152320 Enamine

1-[(3-nitrophenyl)methyl]piperazine dihydrochloride

  • MDL number MFCD04972580
  • Cas Number : 827614-55-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data 2HCl
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-56147

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-56147
Availability details for
Quantity
  • Availability
  • Add to Cart