ENA249152321 Enamine

[3-(4-methanesulfonylphenoxy)propyl](methyl)amine

  • MDL number MFCD12066220
  • Cas Number : 1157381-77-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-56148

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-56148
Availability details for
Quantity
  • Availability
  • Add to Cart