ENA249152327 Enamine

6-cyclopropyl-1-(2-hydroxyethyl)-3-methyl-1H-pyrazolo[3,4-b]pyridine-4-carboxylic acid

  • MDL number MFCD09732511
  • Cas Number : 953729-84-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 184...186
Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-56157

Safety Information

msds https://www.enaminestore.com/catalog/EN300-56157
Availability details for
Quantity
  • Availability
  • Add to Cart