ENA249152328 Enamine

1-(2-bromoethoxy)naphthalene

  • MDL number MFCD02030865
  • Cas Number : 13247-79-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-56159

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-56159
Availability details for
Quantity
  • Availability
  • Add to Cart