ENA249152330 Enamine

{2-amino-1-[4-(trifluoromethyl)phenyl]ethyl}diethylamine

  • MDL number MFCD08059512
  • Cas Number : 914635-97-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-56161

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-56161
Availability details for
Quantity
  • Availability
  • Add to Cart