ENA249152340 Enamine

4-(pyrrolidine-1-carbonyl)-1,2,5-oxadiazol-3-amine

  • MDL number MFCD00456445
  • Cas Number : 302796-45-8
  • Molecular Weight : 182.18

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-56185

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-56185
Availability details for
Quantity
  • Availability
  • Add to Cart