ENA249152341 Enamine

4-(morpholine-4-carbonyl)-1,2,5-oxadiazol-3-amine

  • MDL number MFCD00465017
  • Cas Number : 329206-54-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-56186

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-56186
Availability details for
Quantity
  • Availability
  • Add to Cart