ENA249152342 Enamine

2-methyl-4-(pyrimidin-2-yloxy)aniline

  • MDL number MFCD09049050
  • Cas Number : 926203-60-3
  • Molecular Weight : 201.23

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-56187

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-56187
Availability details for
Quantity
  • Availability
  • Add to Cart