ENA249152346 Enamine

2-butoxypropanoic acid

  • MDL number MFCD11625280
  • Cas Number : 14620-87-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form liquid
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-56194

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-56194
Availability details for
Quantity
  • Availability
  • Add to Cart