ENA249152349 Enamine

2-(5-bromo-2-fluorophenyl)ethan-1-amine

  • MDL number MFCD08448934
  • Cas Number : 615534-62-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-56197

Safety Information

msds https://www.enaminestore.com/catalog/EN300-56197
Availability details for
Quantity
  • Availability
  • Add to Cart