ENA249152351 Enamine

(2-phenylethyl)[(pyridin-4-yl)methyl]amine

  • MDL number MFCD01135003
  • Cas Number : 5713-84-8
  • Molecular Weight : 212.29

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form liquid
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-56199

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-56199
Availability details for
Quantity
  • Availability
  • Add to Cart