ENA249152363 Enamine

1-N-(pyridin-2-ylmethyl)benzene-1,4-diamine

  • MDL number MFCD10694711
  • Cas Number : 408365-68-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 89...91
Purity 95.00
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-56223

Safety Information

msds https://www.enaminestore.com/catalog/EN300-56223
Availability details for
Quantity
  • Availability
  • Add to Cart