ENA249152365 Enamine

1-(1-bromoethyl)-2-nitrobenzene

  • MDL number MFCD11103117
  • Cas Number : 69642-00-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. -20
Shipping Temperature Ice Pack
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-56226

Safety Information

signalword Danger
msds https://www.enaminestore.com/catalog/EN300-56226
pictograms GHS05,GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart