ENA249152366 Enamine

1-(1-bromopropyl)-4-chlorobenzene

  • MDL number MFCD11180266
  • Cas Number : 2940-56-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-56227

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-56227
Availability details for
Quantity
  • Availability
  • Add to Cart