ENA249152368 Enamine

(7-aminoheptyl)dimethylamine

  • MDL number MFCD00047981
  • Cas Number : 22078-09-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-56229

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-56229
Availability details for
Quantity
  • Availability
  • Add to Cart