ENA249152370 Enamine

1,3-diphenylpropan-1-ol

  • MDL number MFCD00266198
  • Cas Number : 14097-24-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form oil
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-56231

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-56231
Availability details for
Quantity
  • Availability
  • Add to Cart