ENA249152379 Enamine

9-oxobicyclo[3.3.1]non-6-ene-3-carboxylic acid

  • Cas Number : 159345-17-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-54569

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-54569
Availability details for
Quantity
  • Availability
  • Add to Cart