ENA249152398 Enamine

4-amino-N-(butan-2-yl)-3-methylbenzamide

  • MDL number MFCD09044603
  • Cas Number : 926233-21-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-56297

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-56297
Availability details for
Quantity
  • Availability
  • Add to Cart