ENA249152410 Enamine

(4-aminobutyl)(methyl)propan-2-ylamine

  • MDL number MFCD09803867
  • Cas Number : 947263-52-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 75...77
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-56313

Safety Information

msds https://www.enaminestore.com/catalog/EN300-56313
Availability details for
Quantity
  • Availability
  • Add to Cart