ENA249152411 Enamine

(Z)-N-[1-(1,3,5-trimethyl-1H-pyrazol-4-yl)ethylidene]hydroxylamine

  • MDL number MFCD13368240
  • Cas Number : 1206252-35-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 162...164
Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-56314

Safety Information

msds https://www.enaminestore.com/catalog/EN300-56314
Availability details for
Quantity
  • Availability
  • Add to Cart