ENA249152413 Enamine

2-bromo-4-fluoro-N,N-dimethylbenzamide

  • MDL number MFCD09927497
  • Cas Number : 1016749-54-4
  • Molecular Weight : 246.08

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-56317

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-56317
Availability details for
Quantity
  • Availability
  • Add to Cart