ENA249152416 Enamine

1-(4-bromophenyl)-5-methyl-1H-1,2,3,4-tetrazole

  • MDL number MFCD07809310
  • Cas Number : 98437-33-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-56340

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-56340
Availability details for
Quantity
  • Availability
  • Add to Cart