ENA249152420 Enamine

1-(2-methoxy-5-methylphenyl)-1H-pyrazole-3-carboxylic acid

  • MDL number MFCD11193612
  • Cas Number : 1152535-19-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-56349

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-56349
Availability details for
Quantity
  • Availability
  • Add to Cart