ENA249152421 Enamine

6-(diethylamino)pyridine-3-carboxylic acid

  • MDL number MFCD07376806
  • Cas Number : 897399-88-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 154...156
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-56350

Safety Information

msds https://www.enaminestore.com/catalog/EN300-56350
Availability details for
Quantity
  • Availability
  • Add to Cart