ENA249152425 Enamine

2-cyclopropyl-1,3-dioxo-2,3-dihydro-1H-isoindole-5-carboxylic acid

  • MDL number MFCD02905236
  • Cas Number : 356576-44-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-56354

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-56354
Availability details for
Quantity
  • Availability
  • Add to Cart