ENA249152435 Enamine

2-(ethylamino)-2-methylpropanenitrile

  • MDL number MFCD10014458
  • Cas Number : 31058-08-9
  • Molecular Weight : 112.17

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. -10
Physical Form liquid
Shipping Temperature Ice Pack
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-56367

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-56367
Availability details for
Quantity
  • Availability
  • Add to Cart