ENA249152439 Enamine

2-amino-1-[2-(trifluoromethyl)phenyl]ethan-1-ol

  • MDL number MFCD08544093
  • Cas Number : 133562-36-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-56379

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-56379
Availability details for
Quantity
  • Availability
  • Add to Cart