ENA249152441 Enamine

3-(aminomethyl)benzamide hydrochloride

  • MDL number MFCD09836255
  • Cas Number : 903556-10-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-56381

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-56381
Availability details for
Quantity
  • Availability
  • Add to Cart