ENA249152442 Enamine

4-hydroxy-N-(pyridin-3-yl)benzamide

  • MDL number MFCD07633133
  • Cas Number : 852980-64-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-56382

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-56382
Availability details for
Quantity
  • Availability
  • Add to Cart