ENA249152444 Enamine

5-(2,4-dimethoxyphenyl)-4H-1,2,4-triazole-3-thiol

  • MDL number MFCD08753809
  • Cas Number : 928710-74-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 250...252
Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-56388

Safety Information

msds https://www.enaminestore.com/catalog/EN300-56388
Availability details for
Quantity
  • Availability
  • Add to Cart