ENA249152446 Enamine

3-(1,3-thiazol-4-yl)propanoic acid

  • MDL number MFCD11180958
  • Cas Number : 933735-27-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 156...158
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-56392

Safety Information

signalword Warning
msds https://www.enaminestore.com/catalog/EN300-56392
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart