ENA249152457 Enamine

2-(prop-2-en-1-yloxy)propanoic acid

  • MDL number MFCD10689722
  • Cas Number : 96692-32-9
  • Molecular Weight : 130.14

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form liquid
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-56406

Safety Information

signalword Danger
pcodes P260,P261,P264,P271,P272,P280,P301+P330+P331,P302+P352,P303+P361+P353,P304+P340,P305+P351+P338,P310,P312,P333+P313,P363,P403+P233,P405,P501
MSDS https://www.enaminestore.com/catalog/EN300-56406
pictograms GHS05,GHS07
hstatements H314,H317,H335
Availability details for
Quantity
  • Availability
  • Add to Cart