ENA249152459 Enamine

4-bromo-N-(2,2,2-trifluoroethyl)aniline

  • MDL number MFCD11153109
  • Cas Number : 50885-29-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-56409

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-56409
Availability details for
Quantity
  • Availability
  • Add to Cart