ENA249152460 Enamine

2-(3,5-dichlorophenyl)ethan-1-amine

  • MDL number MFCD08448789
  • Cas Number : 67851-51-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-56410

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-56410
Availability details for
Quantity
  • Availability
  • Add to Cart