ENA249152465 Enamine

1-(2-cyanoethyl)-5-methyl-1H-pyrazole-4-carboxylic acid

  • MDL number MFCD09051313
  • Cas Number : 1020703-75-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 159...161
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-55404

Safety Information

msds https://www.enaminestore.com/catalog/EN300-55404
Availability details for
Quantity
  • Availability
  • Add to Cart