ENA249152467 Enamine

N'-hydroxy-6-phenoxypyridine-3-carboximidamide

  • Cas Number : 1016730-02-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-56433

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-56433
Availability details for
Quantity
  • Availability
  • Add to Cart