ENA249152469 Enamine

2-(5-chlorothiophen-2-yl)-4-methyl-1,3-thiazole-5-carboxylic acid

  • MDL number MFCD11104838
  • Cas Number : 1152599-58-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-56435

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-56435
Availability details for
Quantity
  • Availability
  • Add to Cart