ENA249152475 Enamine

1-phenyl-1H-pyrazole-4-sulfonyl chloride

  • MDL number MFCD11102951
  • Cas Number : 18336-37-3
  • Molecular Weight : 242.69

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 85-86
Purity 95
Storage temp. +4
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-56445

Safety Information

signalword Danger
pcodes P260,P261,P264,P271,P280,P301+P330+P331,P303+P361+P353,P304+P340,P305+P351+P338,P310,P312,P363,P403+P233,P405,P501
MSDS https://www.enaminestore.com/catalog/EN300-56445
pictograms GHS05,GHS07
hstatements H314,H335
Availability details for
Quantity
  • Availability
  • Add to Cart