ENA249152477 Enamine

3,4-dimethyl-1H-pyrazol-5-amine

  • MDL number MFCD06255247
  • Cas Number : 91159-73-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 63-67
Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-56449

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-56449
Availability details for
Quantity
  • Availability
  • Add to Cart