ENA249152478 Enamine

2-[(pyrrolidin-1-yl)methyl]pyrrolidine

  • MDL number MFCD06653330
  • Cas Number : 195311-28-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-56451

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-56451
Availability details for
Quantity
  • Availability
  • Add to Cart