ENA249152484 Enamine

1-(5-phenyl-1,2,4-oxadiazol-3-yl)methanamine

  • MDL number MFCD07364455
  • Cas Number : 883545-92-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-24436

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-24436
Availability details for
Quantity
  • Availability
  • Add to Cart