ENA249152486 Enamine

N-(furan-2-ylmethyl)-2-(methylamino)acetamide hydrochloride

  • MDL number MFCD13368253
  • Cas Number : 1220035-59-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-56475

Safety Information

msds https://www.enaminestore.com/catalog/EN300-56475
Availability details for
Quantity
  • Availability
  • Add to Cart