ENA249152509 Enamine

1-tert-butyl-1H-1,3-benzodiazol-2-amine

  • MDL number MFCD11202297
  • Cas Number : 35681-38-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-56524

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-56524
Availability details for
Quantity
  • Availability
  • Add to Cart