ENA249152512 Enamine

2-[(1H-indazol-5-yl)formamido]propanoic acid

  • MDL number MFCD11648609
  • Cas Number : 1396973-36-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-56528

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-56528
Availability details for
Quantity
  • Availability
  • Add to Cart