ENA249152517 Enamine

3-phenyl-1-(piperazin-1-yl)propan-1-one

  • MDL number MFCD08442819
  • Cas Number : 117132-90-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-56535

Safety Information

msds https://www.enaminestore.com/catalog/EN300-56535
Availability details for
Quantity
  • Availability
  • Add to Cart